missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-beta-estradiol 97.0+%, TCI America™

Catalog No. M253025MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M253025MG 25 mg
M2530100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M253025MG Supplier TCI America Supplier No. M253025MG
Only null left

Chemical Identifiers

CAS 362-07-2
Molecular Formula C19H26O3
Molecular Weight (g/mol) 302.414
MDL Number MFCD00010489
InChI Key CQOQDQWUFQDJMK-SSTWWWIQSA-N
Synonym 2-methoxyestradiol, panzem, 2-methoxyestradiol-17beta, 2-hydroxyestradol 2-methyl ether, estradiol, 2-methoxy, 2-hydroxyestradiol 2-methyl ether, pulmolar, 2-methoxyestra-1,3,5 10-triene-3,17beta-diol, panzem ncd, 2-methoxy-17beta-estradiol
PubChem CID 66414
ChEBI CHEBI:28955
IUPAC Name (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=CC(=C(C=C34)OC)O

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 25 mg
Formula Weight 302.41
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-beta-estradiol
RTECSNumber KG7537500
TSCA No