missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylphenol 98.0+%, TCI America™

Catalog No. p-7130642
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06245G 5 g
H062425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06245G Supplier TCI America Supplier No. H06245G

Chemical Identifiers

CAS 1195-09-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00040901
InChI Key IFNDEOYXGHGERA-UHFFFAOYSA-N
Synonym isocreosol, 5-methylguaiacol, 6-methoxy-m-cresol, isocreosole, phenol, 2-methoxy-5-methyl, 2-hydroxy-4-methylanisole, m-cresol, 6-methoxy, 2-methoxy-5-methyl-phenol, unii-av5v76ou33, 3-hydroxy-4-methoxytoluene
PubChem CID 14519
IUPAC Name 2-methoxy-5-methylphenol
SMILES CC1=CC(=C(C=C1)OC)O

Specifications

Color White-Yellow
Boiling Point 117°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methoxy-5-methylphenol
RTECSNumber GP1770000
TSCA No