missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzonitrile 97.0+%, TCI America™

Catalog No. M25855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25855G 5 g
M25851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25855G Supplier TCI America Supplier No. M25855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101084-96-2
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.147
MDL Number MFCD02093781
InChI Key MLIKCKXLGYEGAO-UHFFFAOYSA-N
Synonym 2-cyano-5-nitroanisole, acmc-20a4y1, 4-nitro-2-methoxybenzonitrile, 2-methoxy-4-nitro-benzonitrile, 2-methyloxy-4-nitrobenzonitrile, 2-methoxy-4-nitrobenzonitrile
PubChem CID 291867
IUPAC Name 2-methoxy-4-nitrobenzonitrile
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 179°C
Quantity 5 g
UN Number 3439
Formula Weight 178.15
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitrobenzonitrile
EINECSNumber (3)-1825
TSCA No