missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. M186525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M186525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M186525G Supplier TCI America Supplier No. M186525G
Only null left

Chemical Identifiers

CAS 5248-39-5
Molecular Formula C6H10N4O
Molecular Weight (g/mol) 154.173
MDL Number MFCD00585858
InChI Key MNDSUSQBIDHEJU-UHFFFAOYSA-N
PubChem CID 160779
IUPAC Name 4-methoxy-N,6-dimethyl-1,3,5-triazin-2-amine
SMILES CC1=NC(=NC(=N1)OC)NC

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 25 g
Formula Weight 154.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine
RTECSNumber XY2905180
TSCA Yes