missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptothiazoline, 98+%

Catalog No. AAL0492436
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0492436 500 g
AAL0492422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0492436 Supplier Thermo Scientific Chemicals Supplier No. L0492436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-53-7 | C3H5NS2 | 119.2 g/mol

2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 96-53-7
Molecular Formula C3H5NS2
Molecular Weight (g/mol) 119.2
MDL Number MFCD00126013
InChI Key WGJCBBASTRWVJL-UHFFFAOYSA-N
Synonym 2-mercaptothiazoline, 2-thiazolidinethione, 2-thiazoline-2-thiol, thiazolidine-2-thione, metabasal, thyroidan, 2-mercapto-2-thiazoline, mercaptothiazoline, 2-thiothiazolidone, 2-thiazolidenethione
PubChem CID 2723699
IUPAC Name 1,3-thiazolidine-2-thione
SMILES C1CSC(=S)N1

Specifications

Melting Point 103°C to 107°C
Quantity 500 g
UN Number UN2811
Beilstein 106332
Solubility Information Soluble in chloroform,methanol. Slightly soluble in water.
Formula Weight 119.21
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptothiazoline
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-512-1
RTECSNumber XJ6122000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only