missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyrimidine, 98%

Catalog No. AC156740100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156740100 10 g Glass bottle
AC156740500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156740100 Supplier Thermo Scientific Chemicals Supplier No. 156740100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1450-85-7
Molecular Formula C4H4N2S
Molecular Weight (g/mol) 112.15
MDL Number MFCD00006073
InChI Key HBCQSNAFLVXVAY-UHFFFAOYSA-N
Synonym 2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
PubChem CID 1550489
ChEBI CHEBI:53576
IUPAC Name 1H-pyrimidine-2-thione
SMILES C1=CNC(=S)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Formula Weight 112.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Mercaptopyrimidine, 98%
Show More Show Less
EINECSNumber 215-917-3
TSCA TSCA

RUO – Research Use Only