missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, sodium salt hydrate, 98%, Thermo Scientific™

Catalog No. AC255380250
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC255380250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255380250 Supplier Thermo Scientific Chemicals Supplier No. 255380250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 207511-13-5, 3811-73-2
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H4NNaOS·xH2O
Quantity 25g
MDL Number MFCD00151244
Merck Index 10, 7892
Synonym sodium 2-mercaptopyridine n-oxide hydrate
Solubility Information Solubility in water: soluble
InChI Key HTJXGDXCIFMYMU-UHFFFAOYSA-M
SMILES C1=CC=[N+](C(=C1)[S-])[O-].O.[Na+]
IUPAC Name sodium;1-oxidopyridin-1-ium-2-thiolate;hydrate
Molecular Weight (g/mol) 149.15
PubChem CID 43835086
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine-N-oxide,sodium salt hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 223-296-5
RTECSNumber UT9000000
TSCA TSCA

RUO – Research Use Only