missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, sodium salt hydrate, 98%

Catalog No. AC255380050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC255380050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC255380050 Supplier Thermo Scientific Chemicals Supplier No. 255380050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 207511-13-5 | C5H4NNaOS | 149.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207511-13-5 , 3811-73-2
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.14
MDL Number MFCD00151244
InChI Key WNGMMIYXPIAYOB-UHFFFAOYSA-M
Synonym sodium 2-mercaptopyridine n-oxide hydrate
PubChem CID 43835086
IUPAC Name sodium;1-oxidopyridin-1-ium-2-thiolate;hydrate
SMILES [Na+].[O-][N+]1=CC=CC=C1[S-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >260°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Merck Index 10, 7892
Solubility Information Solubility in water: soluble
Formula Weight 149.15
Percent Purity 98%
Physical Form Granular Crystalline Powder
Chemical Name or Material 2-Mercaptopyridine-N-oxide,sodium salt hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 223-296-5
RTECSNumber UT9000000
TSCA TSCA

RUO – Research Use Only