missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptopyridine N-Oxide Sodium Salt Anhydrous 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 287°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 149.14 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Mercaptopyridine N-Oxide Sodium Salt Anhydrous |
Safety and Handling
| EINECSNumber | (9)-1473 |
| RTECSNumber | UT9000000 |
| TSCA | Yes |