missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-Oxide Sodium Salt (40% in Water, ca. 3.3mol/L), TCI America™

Catalog No. M063225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M063225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M063225G Supplier TCI America Supplier No. M063225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A versatile biocide, often used as a preservative in various industrial applications.

Chemical Identifiers

CAS 3811-73-2
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.143
MDL Number MFCD01941547
InChI Key XNRNJIIJLOFJEK-UHFFFAOYSA-N
Synonym sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van
PubChem CID 19658
IUPAC Name sodium;1-oxidopyridine-2-thione
SMILES C1=CC(=S)N(C=C1)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 149.14
Percent Purity 38.0 to 42.0%
Grade Not Available
Physical Form Liquid
Chemical Name or Material 2-Mercaptopyridine N-Oxide Sodium Salt (40% in Water, ca. 3.3mol/L)
EINECSNumber (9)-1473
RTECSNumber UT9000000
TSCA Yes