missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-oxide, 99%

Catalog No. AAA1415222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1415222 100 g
AAA1415206 5 g
AAA1415214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1415222 Supplier Thermo Scientific Chemicals Supplier No. A1415222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1121-31-9 | C5H5NOS | 127.161 g/mol

2-Mercaptopyridine N-oxide is used in ratiometric fluorescence imaging of intracellular zinc ions in mammalian cultures. It is used in the radical coupling of benzoyl derivatives of 2-deoxy-D-ribose, with heterocyclic bases to give C-nucleosides. It acts as a ligand and form coordination complexes with palladium chloride. It is a precursor to O-acyl thiohydroxamates. Further, it serves as an antifungal agent. In addition to this, it is used in the preparation of 1-oxa-2-oxo-3-thia-indolizinium chloride by reaction with carbonyl dichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptopyridine N-oxide is used in ratiometric fluorescence imaging of intracellular zinc ions in mammalian cultures. It is used in the radical coupling of benzoyl derivatives of 2-deoxy-D-ribose, with heterocyclic bases to give C-nucleosides. It acts as a ligand and form coordination complexes with palladium chloride. It is a precursor to O-acyl thiohydroxamates. Further, it serves as an antifungal agent. In addition to this, it is used in the preparation of 1-oxa-2-oxo-3-thia-indolizinium chloride by reaction with carbonyl dichloride.

Solubility
Soluble in tris-hydrochloric acid.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 1121-31-9
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00006196
InChI Key YBBJKCMMCRQZMA-UHFFFAOYSA-N
Synonym pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
PubChem CID 1570
ChEBI CHEBI:36578
IUPAC Name 1-hydroxypyridine-2-thione
SMILES C1=CC(=S)N(C=C1)O

Specifications

Melting Point 69°C to 72°C
Quantity 100 g
Beilstein 906983
Sensitivity Air and light sensitive
Merck Index 14,7994
Solubility Information Soluble in tris-hydrochloric acid.
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material 2-Mercaptopyridine N-oxide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-329-4
RTECSNumber UT8999000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only