missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-nitropyridine 98.0+%, TCI America™

Catalog No. M11025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11025G 5 g
M110225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11025G Supplier TCI America Supplier No. M11025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2127-09-5
Molecular Formula C5H4N2O2S
Molecular Weight (g/mol) 156.16
MDL Number MFCD00030890
InChI Key BHQUBONFIYNJDA-UHFFFAOYSA-N
Synonym 5-Nitro-2-pyridinethiol, 5-Nitro-2-pyridyl Mercaptan
PubChem CID 2763652
IUPAC Name 5-nitro-1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1[N+](=O)[O-]

Specifications

Melting Point 188°C
Quantity 5 g
Formula Weight 156.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-nitropyridine