missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylbenzoic Acid 98.0+%, TCI America™

Catalog No. I09975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09975G 5 g
I09971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09975G Supplier TCI America Supplier No. I09975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2438-04-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00045836
InChI Key BANZVKGLDQDFDV-UHFFFAOYSA-N
PubChem CID 17099
IUPAC Name 2-propan-2-ylbenzoic acid
SMILES CC(C)C1=CC=CC=C1C(=O)O

Specifications

Melting Point 64°C
Color White
Quantity 5 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropylbenzoic Acid
EINECSNumber (3)-1338
TSCA No