missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-3-methoxypyrazine 97.0+%, TCI America™

Catalog No. I03315ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03315ML 5 mL
I03311ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03315ML Supplier TCI America Supplier No. I03315ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24683-00-9
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.22
MDL Number MFCD00006128
InChI Key UXFSPRAGHGMRSQ-UHFFFAOYSA-N
Synonym 2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazine
PubChem CID 32594
IUPAC Name 2-methoxy-3-(2-methylpropyl)pyrazine
SMILES COC1=NC=CN=C1CC(C)C

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 166.22
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-3-methoxypyrazine