missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyrazine 97.0+%, TCI America™

Catalog No. p-7131623
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08371G 1 g
I08375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08371G Supplier TCI America Supplier No. I08371G

Chemical Identifiers

CAS 32111-21-0
Molecular Formula C4H3IN2
Molecular Weight (g/mol) 205.986
MDL Number MFCD01319019
InChI Key OYWPFIUVDKHHGQ-UHFFFAOYSA-N
Synonym iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
PubChem CID 642841
IUPAC Name 2-iodopyrazine
SMILES C1=CN=C(C=N1)I

Specifications

Boiling Point 110°C
Quantity 1 g
Formula Weight 205.99
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodopyrazine