missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stab. with copper

Catalog No. AAA1506614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1506614 25 g
AAA1506622 100 g
AAA1506636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1506614 Supplier Thermo Scientific Chemicals Supplier No. A1506614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 75-30-9 | C3H7I | 169.993 g/mol

2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.993
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Melting Point -90°C
Density 1.701
Boiling Point 88°C to 90°C
Flash Point 42°C (107°F)
Odor Recognizable
Assay Percent Range 98%, stabilized with copper
Refractive Index 1.4982
Quantity 25 g
UN Number UN2392
Beilstein 1098244
Sensitivity Light sensitive
Merck Index 14,5214
Solubility Information Soluble in water(1.4g/L).
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, stabilized with copper
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: IODOPROPANES
EINECSNumber 200-859-3
RTECSNumber TZ4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only