missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. I07705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I07705G Supplier TCI America Supplier No. I07705G
Only null left

Chemical Identifiers

CAS 129112-26-1
Molecular Formula C7H4F3IO3S
Molecular Weight (g/mol) 352.07
MDL Number MFCD07784326
InChI Key AXIDYNSWXPNBLG-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2-Iodophenyl Ester, 2-Iodophenyl Triflate
PubChem CID 10784152
IUPAC Name 2-iodophenyl trifluoromethanesulfonate
SMILES FC(F)(F)S(=O)(=O)OC1=CC=CC=C1I

Specifications

Color Yellow
Quantity 5 g
Formula Weight 352.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodophenyl Trifluoromethanesulfonate