missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 2-Iodophenol | |
| 42°C | |
| C6H5IO | |
| 5 g | |
| KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
| 2-iodophenol | |
| 10784 | |
| 220.01 | |
| Crystalline Powder |
| 533-58-4 | |
| 187°C | |
| MFCD00013963 | |
| o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
| OC1=CC=CC=C1I | |
| 220.01 | |
| CHEBI:16706 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 533-58-4 | |
| 220.01 | |
| KQDJTBPASNJQFQ-UHFFFAOYSA-N | |
| 10784 | |
| 2-iodophenol |
| C6H5IO | |
| MFCD00013963 | |
| o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol | |
| CHEBI:16706 | |
| OC1=CC=CC=C1I |
Safety and Handling
RTECSNumber : SL5500000
TSCA : Yes
Recommended Storage : Refrigerator