accessibility menu, dialog, popup

UserName

OHI

CAS RN 533-58-4

IUPAC Name: 2-iodophenol
Synonyms: 2-iodophenol 2-jodfenol 123i-labeled 2-iodophenol o-jodphenol o-jodfenol 123I-labeled 2-iodophenol 2-iodophenol, 123I-labeled 2-iodophenol, 123i-labeled 2-jodphenol
Molecular Weight (g/mol): 220.01
Molecular Formula: C6H5IO
InChi Key: KQDJTBPASNJQFQ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1I
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)