missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenol 99.0+%, TCI America™

Catalog No. p-7131422
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04605G 5 g
I046025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04605G Supplier TCI America Supplier No. I04605G
Only null left

Chemical Identifiers

CAS 533-58-4
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00013963
InChI Key KQDJTBPASNJQFQ-UHFFFAOYSA-N
Synonym o-iodophenol, phenol, 2-iodo, phenol, o-iodo, o-jodfenol, 2-jodfenol, phenol, iodo, o-jodfenol czech, 2-jodfenol czech, o-jodphenol, 2-iodanylphenol
PubChem CID 10784
ChEBI CHEBI:16706
IUPAC Name 2-iodophenol
SMILES OC1=CC=CC=C1I

Specifications

Melting Point 42°C
Boiling Point 187°C
Quantity 5 g
Formula Weight 220.01
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodophenol
RTECSNumber SL5500000
TSCA Yes
Recommended Storage Refrigerator