missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoethanol, 99%, stab. with copper

Catalog No. AAA1465806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1465806 5 g
AAA1465814 25 g
AAA1465822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1465806 Supplier Thermo Scientific Chemicals Supplier No. A1465806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-76-0 | C2H5IO | 171.97 g/mol

2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoethanol is used in the preparation of the neuroexcitants such as 4-dicarboxylic acid, alfa-kainic acid, 3-(carboxymethyl)pyrrolidine-2 and alfa-isokainic acid.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with acid anhydrides, acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcohol
PubChem CID 12225
IUPAC Name 2-iodoethanol
SMILES OCCI

Specifications

Density 2.202
Boiling Point 85°C (25 mmHg)
Flash Point 65°C (149°F)
Odor Characteristic
Refractive Index 1.572
Linear Formula ICH2CH2OH
Quantity 5 g
UN Number UN2810
Beilstein 1098258
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 171.96
Percent Purity 99%
Chemical Name or Material 2-Iodoethanol, stabilized with copper
Show More Show Less
Hazard Category H227-H300
Hazard Statement GHS H Statement
H300-H310-H341-H227
Fatal if swallowed.
Fatal in contact with skin.
Suspected of causing genetic defects.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P310-P330-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-861-6
RTECSNumber KL3820000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only