missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzotrifluoride (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I038325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I038325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I038325G Supplier TCI America Supplier No. I038325G

Chemical Identifiers

CAS 444-29-1
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00001041
InChI Key IGZGUYVVBABKOY-UHFFFAOYSA-N
Synonym 2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride
PubChem CID 67957
IUPAC Name 1-iodo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)I

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 272.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodobenzotrifluoride (stabilized with Copper chip)