missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacétophénone, 99+%, Thermo Scientific Chemicals

Numéro de catalogue. p-3593257
Modifier l'affichage
Click to view available options
Quantity:
5 g
Packaging:
Bouteille en verre
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC393130050 5 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC393130050 Fournisseur Thermo Scientific Chemicals Code fournisseur 393130050

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de produits d’Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original d’Acros Organics ou la référence SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Identifiants chimiques

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.04
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophényl)éthanone
SMILES CC(=O)C1=CC=CC=C1I

Spécifications

CAS Min % 99.0
CAS Max % 100.0
Color Jaune
Density 1.7460g/mL
Boiling Point 139°C to 140°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Bouteille en verre
Refractive Index 1.617 to 1.619 (20°C, 589nm)
Linear Formula IC6H4C(O)CH3
Quantity 5 g
Specific Gravity 1.746
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 246.04
Percent Purity 99+%
Physical Form Liquide
Chemical Name or Material 2'-Iodoacetophenone
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Utilisation uniquement de la recherche RUO –