missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)aniline 98.0+%, TCI America™

Catalog No. I079425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I079425G 25 g
I07945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I079425G Supplier TCI America Supplier No. I079425G

Chemical Identifiers

CAS 163444-17-5
Molecular Formula C7H5F3IN
Molecular Weight (g/mol) 287.02
MDL Number MFCD02683097
InChI Key UKKWTZPXYIYONW-UHFFFAOYSA-N
Synonym 4-amino-3-iodobenzotrifluoride, 2-iodo-4-trifluoromethyl aniline, 4-trifluoromethyl-2-iodobenzenamine, benzenamine, 2-iodo-4-trifluoromethyl, 2-iodo-4-trifluoromethyl benzenamine, pubchem1632, acmc-209dpp, intermediates-zcf02228, ksc910s3b, 3-iodo-4-aminobenzotrifluoride
PubChem CID 2783322
IUPAC Name 2-iodo-4-(trifluoromethyl)aniline
SMILES NC1=C(I)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 52°C
Quantity 25 g
Formula Weight 287.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)aniline