missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-4-nitrotoluene 97.0+%, TCI America™
Supplier: TCI America I070625G
Specifications
| 2-Iodo-4-nitrotoluene | |
| 52°C | |
| C7H6INO2 | |
| 25 g | |
| BUQSRXQJUZTIEW-UHFFFAOYSA-N | |
| 2-iodo-1-methyl-4-nitrobenzene | |
| 82188 | |
| ≥97.0% (GC) |
| 7745-92-8 | |
| 165°C | |
| MFCD00024329 | |
| 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821 | |
| CC1=CC=C(C=C1I)[N+]([O-])=O | |
| 263.03 | |
| 263.03 | |
| Crystalline Powder |
Chemical Identifiers
| 7745-92-8 | |
| 263.03 | |
| BUQSRXQJUZTIEW-UHFFFAOYSA-N | |
| 82188 | |
| CC1=CC=C(C=C1I)[N+]([O-])=O |
| C7H6INO2 | |
| MFCD00024329 | |
| 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821 | |
| 2-iodo-1-methyl-4-nitrobenzene |
Safety and Handling
TSCA : No