missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-nitrotoluene 97.0+%, TCI America™

Catalog No. p-7131553
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I07065G 5 g
I070625G 25 g
2 options

Catalog No. I07065G

Supplier: TCI America I07065G

Only null left

Chemical Identifiers

CAS 7745-92-8
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD00024329
InChI Key BUQSRXQJUZTIEW-UHFFFAOYSA-N
Synonym 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
PubChem CID 82188
IUPAC Name 2-iodo-1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1I)[N+]([O-])=O

Specifications

Melting Point 52°C
Boiling Point 165°C
Quantity 5 g
Formula Weight 263.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-4-nitrotoluene