accessibility menu, dialog, popup

UserName

CAS RN 7745-92-8

IUPAC Name: 2-iodo-1-methyl-4-nitrobenzene
Synonyms: 2-iodo-1-methyl-4-nitrobenzene
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: BUQSRXQJUZTIEW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1I)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)