missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyterephthalic Acid 98.0+%, TCI America™

Catalog No. H13855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13855G 5 g
H13851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13855G Supplier TCI America Supplier No. H13855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 636-94-2
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD09835368
InChI Key CDOWNLMZVKJRSC-UHFFFAOYSA-N
PubChem CID 97257
IUPAC Name 2-hydroxyterephthalic acid
SMILES C1=CC(=C(C=C1C(=O)O)O)C(=O)O

Specifications

Melting Point 322°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyterephthalic Acid