missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoline, 98%

Catalog No. AC418810100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418810100 10 g Glass bottle
AC418810010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418810100 Supplier Thermo Scientific Chemicals Supplier No. 418810100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59-31-4 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59-31-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006743
InChI Key LISFMEBWQUVKPJ-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone
PubChem CID 6038
ChEBI CHEBI:18289
IUPAC Name 1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 197.0°C to 200.0°C
Color Brown-Purple or Purple to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 1826
Formula Weight 145.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxyquinoline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 200-420-6
RTECSNumber FG7175000
TSCA TSCA

RUO – Research Use Only