missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine 98.0+%, TCI America™

Catalog No. H033025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H033025G 25 g
H0330250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H033025G Supplier TCI America Supplier No. H033025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-08-5 | C5H5NO | 95.10 g/mol

Chemical Identifiers

CAS 142-08-5
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006268
InChI Key UBQKCCHYAOITMY-UHFFFAOYSA-N
Synonym 2-hydroxypyridine, pyridin-2-ol, 2-pyridinol, 2-pyridone, pyridin-2 1h-one, 2 1h-pyridinone, 2-pyridinone, 2-oxopyridine, 2 1h-pyridone, alpha-pyridone
PubChem CID 8871
ChEBI CHEBI:16540
IUPAC Name 1,2-dihydropyridin-2-one
SMILES O=C1NC=CC=C1

Specifications

Melting Point 107°C
Boiling Point 281°C
Quantity 25 g
UN Number 2811
Formula Weight 95.10
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxypyridine
EINECSNumber (5)-0722
RTECSNumber UV1144050
TSCA Yes