missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine-N-oxide, 97%, Thermo Scientific™

Numéro de catalogue. AC243795000
Modifier l'affichage
Click to view available options
Quantité:
25g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC243795000 500g
AC243790250 25g
AC243791000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC243795000 Fournisseur Thermo Scientific Chemicals Code fournisseur 243795000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 13161-30-3
Formule moléculaire C5H5NO2
Poids moléculaire (g/mol) 111.1
Clé InChI SNUSZUYTMHKCPM-UHFFFAOYSA-N
Synonyme 2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
PubChem CID 69975
Nom de l’IUPAC 1-hydroxypyridin-2-one
SOURIRES C1=CC(=O)N(C=C1)O

Spécifications

Nom ou substance chimique 2-Hydroxypyridine-N-oxide
Point d’éclair 100°C
Spectre infrarouge Authentic
Quantité 500g
Poids de formule 111.1
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 236-100-8

RUO – Research Use Only