missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480

Catalog No. AAH3113306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3113306 5 g
AAH3113314 25 g
AAH3113322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3113306 Supplier Thermo Scientific Chemicals Supplier No. H3113306
Only null left

CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol

2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 128446-35-5
Molecular Formula C44H75O36
Molecular Weight (g/mol) 1180.05
MDL Number MFCD00069372

Specifications

Melting Point 305°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight ≈1250 to 1480 d
Chemical Name or Material (2-Hydroxypropyl)-beta-cyclodextrin
EINECSNumber 420-920-1
RTECSNumber GU2293332
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only