missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenylacetic acid, 98%

Catalog No. AAA1392418
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1392418 50 g
AAA1392409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1392418 Supplier Thermo Scientific Chemicals Supplier No. A1392418
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-75-5 | C8H8O3 | 152.149 g/mol

2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

Solubility
Soluble in Methanol and Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 614-75-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004323
InChI Key CCVYRRGZDBSHFU-UHFFFAOYSA-N
Synonym 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid
PubChem CID 11970
ChEBI CHEBI:28478
IUPAC Name 2-(2-hydroxyphenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)O

Specifications

Melting Point 145°C to 147°C
Quantity 50 g
Beilstein 908000
Solubility Information Soluble in Methanol and Water.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-393-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only