missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)pyridine-5-boronic acid, 97%

Catalog No. AAH53319MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53319MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53319MD Supplier Thermo Scientific Chemicals Supplier No. H53319MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 913835-98-0 | C6H8BNO3 | 152.944 g/mol

2-(Hydroxymethyl)pyridine-5-boronic acid is widely employed as intermediate in organic synthesis. It also acts as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Hydroxymethyl)pyridine-5-boronic acid is widely employed as intermediate in organic synthesis. It also acts as a laboratory reagent.

Notes
Store in dry, dark and ventilated place. Incompatible with strong oxidizi

Chemical Identifiers

CAS 913835-98-0
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.944
MDL Number MFCD09027230
InChI Key MSUQKXAJTUAESD-UHFFFAOYSA-N
Synonym 6-hydroxymethyl pyridin-3-yl boronic acid, 6-hydroxymethyl pyridine-3-boronic acid, 2-hydroxymethyl pyridine-5-boronic acid, 6-hydroxymethyl pyridin-3-ylboronic acid, boronicacid, b-6-hydroxymethyl-3-pyridinyl, 5-boronopyridin-2-yl methanol, 6-hydroxymethyl-3-pyridineboronic acid, acmc-209raj, 6-hydroxymethyl-3-pyridyl boronic acid
PubChem CID 44118255
IUPAC Name [6-(hydroxymethyl)pyridin-3-yl]boronic acid
SMILES B(C1=CN=C(C=C1)CO)(O)O

Specifications

Quantity 250 mg
Formula Weight 152.94
Percent Purity 97%
Chemical Name or Material 2-(Hydroxymethyl)pyridine-5-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only