missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H13451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H13451G Supplier TCI America Supplier No. H13451G

Specifications

Chemical Name or Material 2-(Hydroxymethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 87199-14-2
Color White-Yellow
Molecular Formula C7H9BO3
Quantity 1 g
MDL Number MFCD02258966
Synonym 2-hydroxymethylphenylboronic acid, 2-hydroxymethyl phenylboronic acid, 2-hydroxymethyl phenyl boronic acid, 2-hydroxymethyl benzeneboronic acid, boronic acid, 2-hydroxymethyl phenyl, 2-hydroxymethylpheny, acmc-209qil, 2-hydromethylphenylboronicacid, ksc489m2p
InChI Key AFHOBSCDNXGFMO-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1B(O)O
IUPAC Name [2-(hydroxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 151.96
PubChem CID 4374262
Formula Weight 151.96
Physical Form Crystalline Powder
Show More Show Less