missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)-2-nitro-1,3-propanediol 99.0+%, TCI America™

Catalog No. H0271500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0271500G 500 g
H027125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0271500G Supplier TCI America Supplier No. H0271500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126-11-4
Molecular Formula C4H9NO5
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007395
InChI Key OLQJQHSAWMFDJE-UHFFFAOYSA-N
Synonym tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro
PubChem CID 31337
IUPAC Name 2-(hydroxymethyl)-2-nitropropane-1,3-diol
SMILES OCC(CO)(CO)[N+]([O-])=O

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 500 g
Formula Weight 151.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Hydroxymethyl)-2-nitro-1,3-propanediol
EINECSNumber (9)-0900
RTECSNumber TY7350000
TSCA Yes