missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyisobutyric acid, 98%, Thermo Scientific™

Catalog No. p-145373
Change view
Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173100250 25g Plastic bottle
AC173101000 100g Plastic bottle
AC173100010 1kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173100250 Supplier Thermo Scientific Chemicals Supplier No. 173100250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 594-61-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004459
InChI Key BWLBGMIXKSTLSX-UHFFFAOYSA-N
Synonym 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid
PubChem CID 11671
ChEBI CHEBI:50129
IUPAC Name 2-hydroxy-2-methylpropanoic acid
SMILES CC(C)(O)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Boiling Point 84°C (1.5mmHg)
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)2C(OH)CO2H
Quantity 25g
Beilstein 03, 313
Solubility Information Solubility in water: 1900g/L (22°C). Other solubilities: soluble in methanol,diethyl ether and hot benzene,sparingly soluble in cold benzene
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material 2-Hydroxyisobutyric acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 209-848-8
TSCA TSCA

RUO – Research Use Only