missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyindan 98.0+%, TCI America™

Numéro de catalogue. H051725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H051725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H051725G Fournisseur TCI America Code fournisseur H051725G
Il en reste null

Identifiants chimiques

CAS 4254-29-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00003800
InChI Key KMGCKSAIIHOKCX-UHFFFAOYSA-N
Synonym 2-indanol, indan-2-ol, 2-hydroxyindan, 2-hydroxyhydrindene, 2-hydroxyindane, 1h-inden-2-ol, 2,3-dihydro, 2-indanol;, pubchem9652, acmc-1asoy
PubChem CID 77936
IUPAC Name 2,3-dihydro-1H-inden-2-ol
SMILES C1C(CC2=CC=CC=C21)O

Spécifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyindan