missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Starch, Spectrum™ Chemical
Description
Spectrum™ Chemical 2-Hydroxyethyl Starch
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Poly Pail |
| Quantity | 1 kg |
| Grade | Reagent |