missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Starch, Spectrum™ Chemical
Description
Spectrum™ Chemical 2-Hydroxyethyl Starch
Spécifications
Spécifications
| CAS Max % | 100% |
| Packaging | Poly Bottle |
| Quantity | 250 g |
| Grade | Reagent |