missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycarbazole, 97%

Catalog No. AAH2776514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2776514 25 g
AAH2776506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2776514 Supplier Thermo Scientific Chemicals Supplier No. H2776514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-79-3 | C12H9NO | 183.21 g/mol

Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 86-79-3
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00004962
InChI Key GWPGDZPXOZATKL-UHFFFAOYSA-N
Synonym 2-hydroxycarbazole, carbazol-2-ol, ccris 5301, 2-hydroxycarbazol, 7-hydroxycarbazole, 2-hydroxy carbazole, pubchem9945, pubchem9949, 2-hydroxy-9h-carbazol, 2-hydroxy-9h-carbazole
PubChem CID 93551
IUPAC Name 9H-carbazol-2-ol
SMILES OC1=CC2=C(C=C1)C1=C(N2)C=CC=C1

Specifications

Melting Point 262°C to 273°C
Odor Odorless
Quantity 25 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 2-Hydroxycarbazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-699-7
RTECSNumber FE6355000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only