missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzothiazole, 98%

Numéro de catalogue. p-7023264
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1664806 5 g
AAA1664814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1664806 Fournisseur Thermo Scientific Chemicals Code fournisseur A1664806

CAS : 934-34-9 | C7H5NOS | 151,183 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de Alfa Aesar produits. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original Alfa Aesar ou la référence au SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Identifiants chimiques

CAS 934-34-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00022868
InChI Key YEDUAINPPJYDJZ-UHFFFAOYSA-N
Synonym benzothiazolone, 2-hydroxybenzothiazole, 2 3h-benzothiazolone, 2-benzothiazolol, 2-benzothiazolone, 1,3-benzothiazol-2-ol, 2-benzothiazolinone, 3h-benzothiazol-2-one, benzothiazolol, benzo d thiazol-2 3h-one
PubChem CID 13625
ChEBI CHEBI:115196
IUPAC Name 3H-1,3-benzothiazol-2-one
SMILES C1=CC=C2C(=C1)NC(=O)S2

Spécifications

Melting Point 136°C to 140°C
Quantity 5 g
Formula Weight 151.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzothiazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 213-281-1
TSCA Yes
Recommended Storage Température ambiante

RUO – Usage réservé à la recherche