missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzenethiol 97.0+%, TCI America™

Supplier:  TCI America H11775G

 View more versions of this product

Catalog No. H11775G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Hydroxybenzenethiol
6°C
217°C
5 g
2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
C1=CC=C(C(=C1)O)S
126.173
126.17
Liquid
1121-24-0
Yellow
C6H6OS
2922
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
2-sulfanylphenol
70718
≥97.0% (GC)

Chemical Identifiers

1121-24-0
126.173
2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
2-sulfanylphenol
C6H6OS
VMKYTRPNOVFCGZ-UHFFFAOYSA-N
70718
C1=CC=C(C(=C1)O)S
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1096

RTECSNumber : SL7375000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions