accessibility menu, dialog, popup

UserName

CAS RN 1121-24-0

OHHS

CAS RN 1121-24-0

IUPAC Name: 2-sulfanylphenol
Synonyms: O-mercaptophenol
Molecular Weight (g/mol): 126.17
Molecular Formula: C6H6OS
InChi Key: VMKYTRPNOVFCGZ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1S
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)