missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzenethiol 97.0+%, TCI America™

Catalog No. p-7130914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11775G 5 g
H117725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11775G Supplier TCI America Supplier No. H11775G
Only null left

Chemical Identifiers

CAS 1121-24-0
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
InChI Key VMKYTRPNOVFCGZ-UHFFFAOYSA-N
Synonym 2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
PubChem CID 70718
IUPAC Name 2-sulfanylphenol
SMILES C1=CC=C(C(=C1)O)S

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 217°C
Quantity 5 g
UN Number 2922
Formula Weight 126.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxybenzenethiol
EINECSNumber (3)-1096
RTECSNumber SL7375000
TSCA Yes