missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methyl-5-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH2577006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2577006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2577006 Supplier Thermo Scientific Chemicals Supplier No. H2577006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28489-45-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00092010
InChI Key AJDDHNXZBGZBJN-UHFFFAOYSA-N
Synonym 6-methyl-5-nitropyridin-2-ol, 2-hydroxy-6-methyl-5-nitropyridine, 6-hydroxy-3-nitro-2-picoline, 2-hydroxy-5-nitro-6-picoline, 6-hydroxy-2-methyl-3-nitropyridine, 6-methyl-5-nitropyridin-2 1h-one, 2 1h-pyridinone, 6-methyl-5-nitro, 6-methyl-5-nitro-2-pyridinol, 6-methyl-5-nitro-pyridin-2-ol, pubchem10387
PubChem CID 543053
IUPAC Name 6-methyl-5-nitro-1H-pyridin-2-one
SMILES CC1=C(C=CC(=O)N1)[N+](=O)[O-]

Specifications

Melting Point 233°C to 236°C
Quantity 5 g
Formula Weight 154.13
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-6-methyl-5-nitropyridine
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only