missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methoxybenzaldehyde, 98+%

Catalog No. AAL1314503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1314503 1 g
AAL1314506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1314503 Supplier Thermo Scientific Chemicals Supplier No. L1314503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 700-44-7 | C8H8O3 | 152.149 g/mol

2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 700-44-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00151830
InChI Key DZJPDDVDKXHRLF-UHFFFAOYSA-N
PubChem CID 69692
IUPAC Name 2-hydroxy-6-methoxybenzaldehyde
SMILES COC1=CC=CC(=C1C=O)O

Specifications

Melting Point 68°C to 74°C
Quantity 1 g
Beilstein 1101524
Sensitivity Air Sensitive
Synonym 6-methoxysalicylaldehyde, 2-hydroxy-6-methoxy-benzaldehyde, 2-hydroxy-6-methoxybenzal dehyde, 2-hydroxy-6-methoxy benzaldehyde, benzaldehyde, 2-hydroxy-6-methoxy, pubchem8210, 2-methoxy-6-hydroxybenzaldehyde, benzaldehyde,2-hydroxy-6-methoxy, 2-hydroxy-6-methyloxy benzaldehyde
Solubility Information Slightly soluble in water.
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-6-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-844-6
TSCA No

RUO – Research Use Only