missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde 96.0+%, TCI America™

Catalog No. H08815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08815G 5 g
H088125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08815G Supplier TCI America Supplier No. H08815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
EINECSNumber (3)-1200
RTECSNumber BZ2800000
TSCA No