missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%

Catalog No. AC164160050
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164160050 5 g Glass bottle
AC164160100 10 g Glass bottle
AC164160500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164160050 Supplier Thermo Scientific Chemicals Supplier No. 164160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 672-13-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4°C
Color Yellow
Density 1.2100g/mL
Boiling Point 103°C (2.5 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5770 to 1.579
Linear Formula HOC6H3(OCH3)CHO
Quantity 5 g
Beilstein 08, 244
Specific Gravity 1.21
Formula Weight 152.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-589-

RUO – Research Use Only