missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%

Catalog No. AAA1575322
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1575322 100 g
AAA1575306 5 g
AAA1575314 25 g
AAA1575318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1575322 Supplier Thermo Scientific Chemicals Supplier No. A1575322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 672-13-9 | C8H8O3 | 152.149 g/mol

2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O

Specifications

Melting Point 13°C to 16°C
Density 1.221
Boiling Point 248°C to 252°C
Flash Point >110°C (230°F)
Refractive Index 1.578
Quantity 100 g
Beilstein 1100532
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform. Slightly miscible with water.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-589-0
RTECSNumber BZ2800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only